Learn More
Aviva Systems Biology Corp SLC22A5 Antibody - C-terminal region (ARP63404_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63404_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A5. It was validated on Western Blot. . Alternate names: CDSP, FLJ46769, OCTN2, OCTN2VTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-961