Learn More
Aviva Systems Biology Corp SLC22A7 Antibody - N-terminal region (ARP42306_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42306_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC22A7. Alternate names: MGC24091, MGC45202, NLT, OAT2ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-58178