Learn More
Aviva Systems Biology Corp SLC22A7 Antibody - C-terminal region (ARP42708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC22A7. Alternate names: MGC24091, MGC45202, NLT, OAT2ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53350