Learn More
Aviva Systems Biology Corp SLC22A8 Antibody - middle region (ARP41548_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41548_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC22A8. Alternate names: MGC24086, OAT3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit
Catalog No. 50-014-57898