Learn More
Aviva Systems Biology Corp SLC23A2 Antibody - middle region (ARP43902_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43902_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC23A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC23A2. Alternate names: KIAA0238, NBTL1, SLC23A1, SVCT2, YSPL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58663