Learn More
Aviva Systems Biology Corp SLC23A3 Antibody - N-terminal region (ARP44103_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44103_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC23A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC23A3. Alternate names: E2BP3, FLJ31168, SVCT3, Yspl1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58718