Learn More
Aviva Systems Biology Corp SLC25A11 Antibody - N-terminal region (ARP43849_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43849_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A11. Alternate names: OGC, SLC20A4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58646