Learn More
Aviva Systems Biology Corp SLC25A12 Antibody - middle region (ARP43858_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43858_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC25A12. Alternate names: ARALAR, ARALAR1, AGC1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58650