Learn More
Aviva Systems Biology Corp SLC25A14 Antibody - N-terminal region (ARP43861_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43861_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A14. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A14. Alternate names: UCP5, BMCP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-53380