Learn More
Aviva Systems Biology Corp Slc25a15 Antibody - N-terminal region (ARP43946_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43946_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc25a15. It was validated on Western Blot. . Alternate names: D630044L02Rik, Ornt1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-112