Learn More
Aviva Systems Biology Corp SLC25A16 Antibody - N-terminal region (ARP44125_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44125_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A16. Alternate names: D10S105E, GDA, GDC, HGT.1, MGC39851, ML7, hML7ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53405