Learn More
Aviva Systems Biology Corp SLC25A16 Antibody - middle region (ARP44126_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44126_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC25A16. Alternate names: D10S105E, GDA, GDC, HGT.1, MGC39851, ML7, hML7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58725