Learn More
Aviva Systems Biology Corp SLC25A19 Antibody - middle region (ARP44023_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44023_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A19. It was validated on Western Blot. SLC25A19,solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19,DNC; TPC; MUP1; MCPHA; SLC25A19,mitochondrial thiamine pyrophosphate carrier,mitochondrial thiamine pyrophosphate carrier,mitochondrial uncoupling protein 1,solute carrier family 25 (mitochondrial deoxynucleotide carrier), member 19The immunogen is a synthetic peptide directed towards the middle region of human SLC25A19. Alternate names: DNC, MCPHA, MUP1, TPC, THMD3, THMD4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-285-119