Learn More
Aviva Systems Biology Corp SLC25A2 Antibody - C-terminal region (ARP65709_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65709_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human SLC25A2. Alternate names: MGC119151, MGC119153, ORC2, ORNT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-288-547