Learn More
Aviva Systems Biology Corp SLC25A21 Antibody - middle region (ARP44046_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44046_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC25A21. Alternate names: MGC126570, ODC, ODC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58702