Learn More
Aviva Systems Biology Corp SLC25A22 Antibody - N-terminal region (ARP44041_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44041_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A22. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A22. Alternate names: FLJ13044, GC1, EIEE3, NET44ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-58699