Learn More
ABclonal Technology SLC25A23 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143521000UL
Encompass_Preferred
Predicted to enable ATP transmembrane transporter activity. Involved in calcium import into the mitochondrion, positive regulation of mitochondrial calcium ion concentration, and regulation of cellular hyperosmotic salinity response. Located in mitochondrion.
Catalog No. 50-001-08439