Learn More
Aviva Systems Biology Corp SLC25A24 Antibody - middle region (ARP43937_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43937_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC25A24. Alternate names: RP11-356N1.3, APC1, DKFZp586G0123, SCAMC-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58670