Learn More
Aviva Systems Biology Corp SLC25A25 Antibody - N-terminal region (ARP43763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A25. Alternate names: KIAA1896, MCSC, MGC105138, MGC119514, MGC119515, MGC119516, MGC119517, PCSCL, RP11-395P17.4, SCAMC-2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58622