Learn More
Aviva Systems Biology Corp Slc25a27 Antibody - C-terminal region (ARP43880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc25a27. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 3632410G24Rik, 9430092A03Rik, D530043E16Rik, Ucp4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-105