Learn More
Aviva Systems Biology Corp SLC25A28 Antibody - C-terminal region (ARP44054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SLC25A28. Alternate names: DKFZp547C109, MFRN2, MRS3/4, MRS4L, NPD016ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53398