Learn More
Aviva Systems Biology Corp SLC25A29 Antibody - C-terminal region (ARP43788_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43788_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A29. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SLC25A29. Alternate names: C14orf69, FLJ38975, CACLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58630