Learn More
Aviva Systems Biology Corp Slc25a31 Antibody - middle region (ARP53806_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53806_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc25a31. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1700034J06Rik, Ant4, SfecApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-890