Learn More
Aviva Systems Biology Corp SLC25A31 Antibody - N-terminal region (ARP53805_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A31. Alternate names: AAC4, ANT4, DKFZp434N1235, SFEC35kDaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61134