Learn More
Aviva Systems Biology Corp SLC25A32 Antibody - middle region (ARP44050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A32. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC25A32. Alternate names: FLJ23872, MFT, MFTCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58703