Learn More
Aviva Systems Biology Corp SLC25A33 Antibody - middle region (ARP60511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A33. It was validated on Western Blot. . Alternate names: BMSC-MCP, MGC4399ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-287-068