Learn More
Aviva Systems Biology Corp SLC25A35 Antibody - N-terminal region (ARP44177_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44177_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A35. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A35. Alternate names: FLJ40217, MGC120446, MGC120448ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58739