Learn More
Aviva Systems Biology Corp SLC25A36 Antibody - N-terminal region (ARP43984_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43984_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A36. Alternate names: DKFZp564C053, FLJ10618ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58686