Learn More
Aviva Systems Biology Corp SLC25A38 Antibody - middle region (ARP43978_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43978_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A38. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC25A38. Alternate names: FLJ20551, FLJ22703ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58683