Learn More
Aviva Systems Biology Corp SLC25A4 Antibody - N-terminal region (ARP43818_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43818_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A4. Alternate names: ANT, ANT1, PEO2, PEO3, T1, AAC1ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62948