Learn More
Aviva Systems Biology Corp SLC25A44 Antibody - middle region (ARP43955_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43955_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A44. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SLC25A44. Alternate names: FLJ90431, KIAA0446, RP11-54H19.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-114