Learn More
Aviva Systems Biology Corp SLC25A46 Antibody - N-terminal region (ARP44094_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44094_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A46. Alternate names: ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58715