Learn More
Aviva Systems Biology Corp SLC25A46 Antibody - C-terminal region (ARP44095_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44095_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SLC25A46. Alternate names: -ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58716