Learn More
Aviva Systems Biology Corp SLC25A48 Antibody - C-terminal region (ARP52846_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52846_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC153328. It was validated on Western Blot. SLC25A48,solute carrier family 25, member 48,HDMCP; SLC25A48,solute carrier family 25 member 48,solute carrier family 25 member 48,mitochondrial carrier protein-like,HCC-down-regulated mitochondrial carrier proteinThe immunogen is a synthetic peptide directed towards the c terminal region of human LOC153328. Alternate names: HDMCPApplicationWB. Species reactivity: Human
Catalog No. 50-285-774