Learn More
Aviva Systems Biology Corp SLC25A6 Antibody - N-terminal region (ARP58528_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58528_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC25A6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC25A6. Alternate names: ANT3, ANT3Y, MGC17525, AAC3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62344