Learn More
Aviva Systems Biology Corp SLC26A1 Antibody - middle region (ARP44029_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC26A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC26A1. Alternate names: EDM4, SAT-1, SAT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-014-62992