Learn More
Aviva Systems Biology Corp SLC26A1 Antibody - C-terminal region (ARP44028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC26A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC26A1. Alternate names: EDM4, SAT-1, SAT1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58695