Learn More
ABclonal Technology SLC26A2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14561500UL
Encompass_Preferred
The diastrophic dysplasia sulfate transporter is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias. It apparently is critical in cartilage for sulfation of proteoglycans and matrix organization.
Catalog No. 50-001-07073