Learn More
Aviva Systems Biology Corp SLC26A4 Antibody - middle region (ARP41739_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41739_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC26A4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC26A4. Alternate names: DFNB4, PDS, EVA, TDH2BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57974