Learn More
Aviva Systems Biology Corp SLC26A5 Antibody - middle region (ARP44176_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44176_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC26A5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC26A5. Alternate names: DFNB61, MGC118886, MGC118887, MGC118888, MGC118889, PRESApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58738