Learn More
Aviva Systems Biology Corp SLC26A6 Antibody - C-terminal region (ARP61481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC26A6. It was validated on Western Blot. . Alternate names: DKFZp586E1422ApplicationWB. Species reactivity: Human
Catalog No. 50-287-393