Learn More
Aviva Systems Biology Corp SLC27A2 Antibody - N-terminal region (ARP43855_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43855_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC27A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC27A2. Alternate names: ACSVL1, FACVL1, FATP2, HsT17226, VLACS, VLCS, hFACVL1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58649