Learn More
Aviva Systems Biology Corp SLC27A5 Antibody - middle region (ARP41592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC27A5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC27A5. Alternate names: ACSB, ACSVL6, FACVL3, FATP5, FLJ22987, VLACSR, VLCS-H2, VLCSH2, BAL, BACS, FATP-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57919