Learn More
Aviva Systems Biology Corp SLC27A6 Antibody - middle region (ARP43781_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43781_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC27A6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC27A6. Alternate names: ACSVL2, DKFZp779M0564, FACVL2, FATP6, VLCS-H1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58628