Learn More
Aviva Systems Biology Corp SLC2A10 Antibody - middle region (ARP44048_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44048_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC2A10. Alternate names: GLUT10, ATSApplicationWB. Species reactivity: Human
Catalog No. 50-014-53396