Learn More
Aviva Systems Biology Corp Slc2a12 Antibody - C-terminal region (ARP37590_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc2a12. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: GLUT-12, Glut12ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-662