Learn More
Aviva Systems Biology Corp SLC2A12 Antibody - middle region (ARP44105_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44105_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC2A12. Alternate names: GLUT12, GLUT8ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53869