Learn More
Aviva Systems Biology Corp SLC2A2 Antibody - N-terminal region (ARP41706_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41706_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC2A2. Alternate names: GLUT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-57962