Learn More
Aviva Systems Biology Corp SLC2A3 Antibody - middle region (ARP63350_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63350_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A3. It was validated on Western Blot. . Alternate names: FLJ90380, GLUT3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-945