Learn More
Aviva Systems Biology Corp SLC2A4RG Antibody - middle region (ARP33168_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33168_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A4RG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC2A4RG. Alternate names: GEF, HDBP1, Si-1-2, Si-1-2-19, HDBP-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rat
Catalog No. 50-014-54904